MSD GOLD 96-WELL STREPTAVIDIN SECTOR PLATE PACK 30 PLATES 1 * 30 items
Catalog # MESOL15SA-5
Supplier: MESO SCALE DISCOVERY
MSD GOLD 96-WELL STREPTAVIDIN SECTOR PLATE PACK 30 PLATES 1 * 30 items
Catalog # MESOL15SA-5
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, Low-Binding
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
Diluent 12 1 * 200 mL
Supplier: MESO SCALE DISCOVERY
MSD GOLD SULFO-TAG NHS-ESTER CONJUGATION 1 * 2 ST
Supplier: MESO SCALE DISCOVERY
WASH BUFFER (20X
Supplier: MESO SCALE DISCOVERY
MULTI-ARRAY 96 STANDARD PLATE PACK 100 PLATE 1 * 100 items
Supplier: MESO SCALE DISCOVERY