Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

High-resolution buffer, 7,000, -, —, 25 °C, Plastic bottle, 500 mL
Catalog # REGEH70525
Supplier:   REAGECON
Buffer solutions
Catalog # REGEH70525
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

High-resolution buffer, 7,000, -, —, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
47,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
  • Description:
    High-resolution buffer
  • pH accuracy:
    -
  • pH value:
    7,000

Specifications