ContactUsLinkComponent
33 COMPOSES A 100 MG/L CHAQUE DANS HNO3 5%,HF 1% Ag Al As B Ba Be Bi Ca Cd Co Cr Cu Fe Mg Mn Mo Ni Pb Sb Se Si Sn Sr Te Ti Tl V W Zn Zr K Li Na 1 * 500 mL
Catalog # CMPL9029.K1.5N1F.L
33 COMPOSES A 100 MG/L CHAQUE DANS HNO3 5%,HF 1% Ag Al As B Ba Be Bi Ca Cd Co Cr Cu Fe Mg Mn Mo Ni Pb Sb Se Si Sn Sr Te Ti Tl V W Zn Zr K Li Na 1 * 500 mL
Catalog # CMPL9029.K1.5N1F.L
Frequently Bought Together