Stoppers, self realasing, PTFE
34/35, 70 mm
Catalog # COWI010.1134
Supplier: COWIE
Jump to:
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specifications
- NS:34/35
- Height:70 mm