230V, N2: 30L/min, >98%. 0.2L/min, <10ppm. 7 bar g (101.5 psi g), without compressor Agilent 6460/6510/6520/6530 1 * 1 items
Catalog # DHUNLCMS65-0-E
Supplier: DOMNICK HUNTER
230V, N2: 30L/min, >98%. 0.2L/min, <10ppm. 7 bar g (101.5 psi g), without compressor Agilent 6460/6510/6520/6530 1 * 1 items
Catalog # DHUNLCMS65-0-E
Supplier: DOMNICK HUNTER
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK