Sample concentrators, Reacti Vap™
Reacti-Vap III sample concentrator (27 port)
Catalog # HYPETS-18826
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Reacti-Vap Evaporators (9 port)
These evaporators are precision machined gassing manifolds. They provide simple, efficient evaporation by allowing the simultaneous or separate delivery of non-reactive pressurised gas to samples. They easily connect to single block Reacti-Therm™ modules.
Reacti-Vap III Evaporators (27 port)
These evaporators triple the number of samples to evaporate. Nine needles attach to each of the three individually regulated chambers. They easily connect to Reacti- Therm™ III modules.
Specifications
- Description:Reacti-Vap III sample concentrator (27 port)
- Seq:002
Accessories
Product Information
- Catalog #AvailabilityPrice
- Metal rod
Product Information
- Catalog #AvailabilityPrice
- Replacement height adjustment knobs
Product Information
- Catalog #AvailabilityPrice
- Luer-lok fitting
Product Information
- Catalog #AvailabilityPrice
- Replacement tube kit