Orders LinkContactUsLinkComponent
Sample concentrators, Reacti Vap™
Reacti-Vap sample concentrator (9 port)
Catalog # HYPETS-18825
Sample concentrators, Reacti Vap™
Catalog # HYPETS-18825
Sample concentrators, Reacti Vap™
Reacti-Vap sample concentrator (9 port)
Price
1 240,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Reacti-Vap Evaporators (9 port)
These evaporators are precision machined gassing manifolds. They provide simple, efficient evaporation by allowing the simultaneous or separate delivery of non-reactive pressurised gas to samples. They easily connect to single block Reacti-Therm™ modules.
Reacti-Vap III Evaporators (27 port)
These evaporators triple the number of samples to evaporate. Nine needles attach to each of the three individually regulated chambers. They easily connect to Reacti- Therm™ III modules.
Specifications
  • Description:
    Reacti-Vap sample concentrator (9 port)
  • Seq:
    001
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Metal rod
    Metal rod
    HYPETS-18831
    Log in for availability
    1 * 1 ST - 70,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement height adjustment knobs
    Replacement height adjustment knobs
    HYPETS-18829
    Log in for availability
    1 * 1 ST - 37,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Luer-lok fitting
    Luer-lok fitting
    HYPETS-18827
    Log in for availability
    1 * 1 ST - 54,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement tube kit
    Replacement tube kit
    HYPETS-18782
    Log in for availability
    1 * 9 ST - 222,00 €