Order Entry
Belgium
Orders LinkContactUsLinkComponent
Aluminium blocks, Reacti-Block™

Aluminium blocks, Reacti-Block™

Reacti-Block J, — mm, Custom drilling (no holes), —, 76 mm, 94×75×76 mm
Catalog # HYPETS-18809
Aluminium blocks, Reacti-Block™
Catalog # HYPETS-18809
Aluminium blocks, Reacti-Block™
Aluminium blocks, Reacti-Block™

Reacti-Block J, — mm, Custom drilling (no holes), —, 76 mm, 94×75×76 mm

Price
208,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Reacti-Block™ aluminium blocks are available with many hole configurations, machine-drilled to accommodate almost any size Reacti-Vial™ small reaction vial, test tube or microcentrifuge tube.

  • Made of aluminium alloy for optimal thermal conductivity
  • Block should be fit with sample container for proper heat transfer

Each block contains a thermometer well with size 7,1×36,5 mm (Ø×D mm), except blank blocks J and K.

Aluminium block models A-1, C-1, Z-1, M-1, S-1, U-1 and V-1 can be used with all Reacti-Therm™ modules, including those equipped with Reacti-Vap™ evaporators. Blocks B-1 and T-1 are specifically designed for use with Reacti-Vap™ units.

Aluminium block models F, G, J, K and L are designed to be used exclusively with the Reacti-Therm™ modules. These are not compatible with Reacti-Vap™ evaporators.

Specifications
  • No. of Holes:
  • Description:
    Reacti-Block J
  • Seq:
    13
  • W×D×H:
    94×75×76 mm
  • Depth:
    76 mm
  • Well size:
    — mm
  • For:
    Custom drilling (no holes)