ContactUsLinkComponent
VERTICAL PROGRAMMABLE, MULTI-FUNCTION ROTATOR, SUPPLIED WITH STANDARD PLATFORM ACCOMMODATING 26 X 2-15ML TUBES OF UP TO 15MM DIAMETER, 12V POWER SUPPLY 1 * 1 items
Catalog # 444-0723
Supplier:  GRANT INSTRUMENTS
undefined
VERTICAL PROGRAMMABLE, MULTI-FUNCTION ROTATOR, SUPPLIED WITH STANDARD PLATFORM ACCOMMODATING 26 X 2-15ML TUBES OF UP TO 15MM DIAMETER, 12V POWER SUPPLY 1 * 1 items
Catalog # 444-0723
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together