Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette tips

Pipette tips

Type J, 20 µl, -, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic, 1000/bag, 10000/case
  SARS3021
Marketsource
 :   SARSTEDT
Pipette tips
  SARS3021
 :  SARSTEDT
Marketsource
Pipette tips

Type J, 20 µl, -, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic, 1000/bag, 10000/case

 
 
   
1 * 10.000 ST
 

These high quality tips are suitable for the most common pipettes.

  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems

Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.

 
  • Packed:
    1000/bag, 10000/case
  • Sterile:
    -
  • Volume:
    20 µl
  • Seq:
    22
  • Tip style:
    Type J
  • Tip colour:
    Transparent
  • For use with:
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
Accessories

 

  •  
    Description
    Colour
     
     
  • SARS95.1298.001
    Tip system box for pipette tip trays, with hinged lid, empty
    Transparent
       
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.