Safety baskets
Basket with handle, PP, blue, 430×310×220 mm
Catalog # SEMA4953
Supplier: SEMADENI
Aller à :
Product Details & Documents
PP, coloured.
- Anti-static
- Space-saving, stackable
- Good chemical and temperature resistance
The antistatic safety baskets with perforated walls and base comes with collapsible plastic mounting bracket with lock in vertical position.
Spécifications
- Description:Basket with handle, PP, blue
- Seq:03
- W×D×H:430×310×220 mm