Packaging and Storage Boxes
Transparent, 50×48×13 mm, PS
Catalog # SEMA4560
Supplier: SEMADENI
Aller à :
Product Details & Documents
These boxes made of polystyrene (PS) are crystal clear with a good closing tight fitting lid.
- Clear for easy product identification
- The lids are either mounted or not mounted
- Stackable
Spécifications
- Colour:Transparent
- Material:PS
- Seq:03
- W×D×H:50×48×13 mm