Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue forceps, slim 1×2 teeth

Tissue forceps, slim 1×2 teeth

Stainless steel, 120 mm, 1×2 teeth, Straight, 1,7×1,3 mm
N° de catalogue FSCI11023-12
Marketsource
Fournisseur:   FINE SCIENCE TOOLS
Tissue forceps, slim 1×2 teeth
N° de catalogue FSCI11023-12
Fournisseur:  FINE SCIENCE TOOLS
Marketsource
Tissue forceps, slim 1×2 teeth
Tissue forceps, slim 1×2 teeth

Stainless steel, 120 mm, 1×2 teeth, Straight, 1,7×1,3 mm

Prix
20,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Length:
    120 mm
  • Material:
    Stainless steel
  • Seq:
    02
  • Tip style:
    1×2 teeth
  • Tip size:
    1,7×1,3 mm
  • Tip shape:
    Straight