Order Entry
Belgium
Orders LinkContactUsLinkComponent
Wagner scissors

Wagner scissors

Sharp/sharp, curved tips, 120 mm, 30 mm, +
  FSCI14071-12
Marketsource
Wagner scissors
  FSCI14071-12
Marketsource
Wagner scissors
Wagner scissors

Sharp/sharp, curved tips, 120 mm, 30 mm, +

 
72,50 €
   
1 * 1 Pce
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/sharp, curved tips
  • Length:
    120 mm
  • Seq:
    03
  • Serrated:
    +
  • Blade length:
    30 mm