Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

S&T forceps, SuperGrip Tips

JF-5TC, Inox, 110 mm, Supergrip, Straight, 0,3×0,25 mm
Catalog # FSCI00632-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
S&T forceps, SuperGrip Tips
Catalog # FSCI00632-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
S&T forceps, SuperGrip Tips
S&T forceps, SuperGrip Tips

JF-5TC, Inox, 110 mm, Supergrip, Straight, 0,3×0,25 mm

Price
270,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

SuperGrip tips are processed with MicroCrystals®, a method developed by S&T to infuse tungsten carbide on stainless steel. The formation of this ultra-fine coating guarantees atraumatic handling of tissue while improving the grip dramatically. The gripping surface is similar to a match head.

Recommended when forceps are utilized as needle holders or when slippery tissues might be encountered.

Specifications
  • Type:
    JF-5TC
  • Length:
    110 mm
  • Material:
    Inox
  • Seq:
    02
  • Tip style:
    Supergrip
  • Tip size:
    0,3×0,25 mm
  • Tip shape:
    Straight