Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Fine scissors

Sharp/sharp, straight tips, Ceramacut®, 115 mm, 25 mm, +
  FSCI14958-11
Marketsource
Fine scissors
  FSCI14958-11
Marketsource
Fine scissors
Fine scissors

Sharp/sharp, straight tips, Ceramacut®, 115 mm, 25 mm, +

 
251,50 €
   
1 * 1 ST
 

These scissors are made of ceramic coated stainless steel.

 
  • Type:
    Sharp/sharp, straight tips, Ceramacut®
  • Length:
    115 mm
  • Seq:
    03
  • Serrated:
    +
  • Blade length:
    25 mm