Order Entry
Belgium
Orders LinkContactUsLinkComponent
Surgical scissors, sharp-blunt

Surgical scissors, sharp-blunt

Sharp/blunt, curved tips, 145 mm, 47 mm
Catalog # FSCI14004-14
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, sharp-blunt
Catalog # FSCI14004-14
Supplier:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, sharp-blunt
Surgical scissors, sharp-blunt

Sharp/blunt, curved tips, 145 mm, 47 mm

Price
39,50 €
Log in for availability
1 * 1 Pce
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/blunt, curved tips
  • Length:
    145 mm
  • Blade length:
    47 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.