Order Entry
Belgium
Orders LinkContactUsLinkComponent
Surgical scissors, sharp

Surgical scissors, sharp

Sharp/sharp, straight tips, 120 mm, 41 mm
Catalog # FSCI14002-12
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, sharp
Catalog # FSCI14002-12
Supplier:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, sharp
Surgical scissors, sharp

Sharp/sharp, straight tips, 120 mm, 41 mm

Price
33,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/sharp, straight tips
  • Length:
    120 mm
  • Seq:
    02
  • Blade length:
    41 mm