Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Surgical scissors, sharp-blunt

Sharp/blunt, straight tips, 130 mm, 42 mm
Catalog # FSCI14001-13
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, sharp-blunt
Catalog # FSCI14001-13
Supplier:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, sharp-blunt
Surgical scissors, sharp-blunt

Sharp/blunt, straight tips, 130 mm, 42 mm

Price
32,00 €
Connexion Pour la disponibilité
1 * 1 Pce
Product Details & Documents

These scissors are made of stainless steel.

Spécifications
  • Type:
    Sharp/blunt, straight tips
  • Length:
    130 mm
  • Seq:
    03
  • Blade length:
    42 mm