Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Surgical scissors, sharp

Sharp/sharp, curved tips, 130 mm, 42 mm
  FSCI14005-13
Marketsource
Surgical scissors, sharp
  FSCI14005-13
Marketsource
Surgical scissors, sharp
Surgical scissors, sharp

Sharp/sharp, curved tips, 130 mm, 42 mm

 
39,00 €
   
1 * 1 Pce
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/sharp, curved tips
  • Length:
    130 mm
  • Seq:
    03
  • Blade length:
    42 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.