Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Surgical scissors, sharp-blunt

Sharp/blunt, straight tips, 165 mm, 53 mm
  FSCI14001-16
Marketsource
Surgical scissors, sharp-blunt
  FSCI14001-16
Marketsource
Surgical scissors, sharp-blunt
Surgical scissors, sharp-blunt

Sharp/blunt, straight tips, 165 mm, 53 mm

 
36,00 €
   
1 * 1 ST
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/blunt, straight tips
  • Length:
    165 mm
  • Seq:
    08
  • Blade length:
    53 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.