Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fine scissors, large loops

Fine scissors, large loops

Sharp/sharp, curved tips, large loops, 100 mm, 25 mm
  FSCI14041-10
Marketsource
Fine scissors, large loops
  FSCI14041-10
Marketsource
Fine scissors, large loops
Fine scissors, large loops

Sharp/sharp, curved tips, large loops, 100 mm, 25 mm

 
73,50 €
   
1 * 1 Pce
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/sharp, curved tips, large loops
  • Length:
    100 mm
  • Seq:
    01
  • Blade length:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.