Mayo-Noble scissors
Blunt/blunt, straight tips, 165 mm, 37 mm
FSCI14014-17
Marketsource
These scissors are made of stainless steel.
- Type:Blunt/blunt, straight tips
- Length:165 mm
- Seq:02
- Blade length:37 mm
These scissors are made of stainless steel.