Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Mathieu needle holders

Clamping length of 14 mm, Lockable, extra narrow profile, Stainless steel, 140 mm, Serrated, Straight, 2,3 mm
  FSCI12010-14
Marketsource
Mathieu needle holders
  FSCI12010-14
Marketsource
Mathieu needle holders
Mathieu needle holders

Clamping length of 14 mm, Lockable, extra narrow profile, Stainless steel, 140 mm, Serrated, Straight, 2,3 mm

 
82,50 €
   
1 * 1 Pce
 
 
  • Type:
    Lockable, extra narrow profile
  • Description:
    Clamping length of 14 mm
  • Length:
    140 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip style:
    Serrated
  • Tip width:
    2,3 mm
  • Tip shape:
    Straight