Order Entry
Belgium
Orders LinkContactUsLinkComponent
Moria Iris forceps

Moria Iris forceps

MC31B, Inox, 100 mm, Smooth, Curved, 0,5×,04 mm, 0,5 mm
Catalog # FSCI11373-12
Marketsource
Supplier:   FINE SCIENCE TOOLS
Moria Iris forceps
Catalog # FSCI11373-12
Supplier:  FINE SCIENCE TOOLS
Marketsource
Moria Iris forceps
Moria Iris forceps

MC31B, Inox, 100 mm, Smooth, Curved, 0,5×,04 mm, 0,5 mm

Price
145,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    MC31B
  • Length:
    100 mm
  • Material:
    Inox
  • Seq:
    03
  • Tip style:
    Smooth
  • Tip size:
    0,5×,04 mm
  • Tip width:
    0,5 mm
  • Tip shape:
    Curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.