Coagulation tubes, BD Vacutainer®
Blood Collection tube, BD Vacutainer®, Plastic, BD Hemogard™, buffered Sodium citrate (0,129 M), 75 mm, 13 mm, Light blue, 1,8 ml
Catalog # BDAM363093UK
Jump to:
Discontinued
Product Details & Documents
PET/PP or glass.
- With buffered citrate
- With paper lable
- Hemogard™ closure
Citrate tubes
Specifications
- Closure type:BD Hemogard™
- Ø:13 mm
- Volume:1,8 ml
- Description:Blood Collection tube, BD Vacutainer®
- Length:75 mm
- Material:Plastic
- Cap colour:Light blue
- Additive:buffered Sodium citrate (0,129 M)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



