Order Entry
Belgium
Orders LinkContactUsLinkComponent
Plasmid Maxiprep kit, GeneJET™

Plasmid Maxiprep kit, GeneJET™

GeneJET™ Plasmid Maxiprep Kit, 10 preps, 1 KIT
N° de catalogue K0491
Fournisseur:   Thermo Fisher Scientific
Plasmid Maxiprep kit, GeneJET™
N° de catalogue K0491
Numéro CAS:  
Plasmid Maxiprep kit, GeneJET™
Plasmid Maxiprep kit, GeneJET™

GeneJET™ Plasmid Maxiprep Kit, 10 preps, 1 KIT

Numéro CAS:  
Prix
190,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents

GeneJET™ Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. GeneJET™ Plasmid Maxiprep Kit utilises silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 µg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

  • Efficient: High yields up to 750 µg of high quality plasmid DNA from 250 ml of bacterial culture
  • High concentration of DNA purified: >400 ng/µl
  • Convenient: Spin column format, centrifuge or vacuum protocols
  • Fast: Procedure takes 60 minutes, considerably faster than using gravity flow columns

Livraison: Each kit includes resuspension solution, lysis solution, neutralisation solution, endotoxin binding reagent, Wash Solution I (concentrated), Wash Solution II (concentrated), RNase A solution, elution buffer (10 mM Tris-HCl, pH 8,5), GeneJET™ Maxi purification columns pre-assembled with collection tubes (50 ml), and collection tubes (50 ml).

Spécifications

Spécifications

  • Pk:
    1 KIT
  • Description:
    GeneJET™ Plasmid Maxiprep Kit, 10 preps
  • Cat. No.:
    K0491
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Description:
    GeneJET™ Plasmid Maxiprep Kit
  • Sample type:
    Bacteria

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.