Order Entry
Belgium
Orders LinkContactUsLinkComponent
Reverse transcriptase, Maxima™

Reverse transcriptase, Maxima™

Maxima™ Reverse Transcriptase, 10 000 units (200 U/µl), 1 KIT
Catalog # EP0742
Reverse transcriptase, Maxima™
Catalog # EP0742
CAS Number:  
Reverse transcriptase, Maxima™
Reverse transcriptase, Maxima™

Maxima™ Reverse Transcriptase, 10 000 units (200 U/µl), 1 KIT

CAS Number:  
Price
276,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Maxima™ Reverse Transcriptase is an advanced enzyme derived by in vitro evolution of M-MuLV RT for RT-qPCR application. The enzyme features high thermostability, robustness and increased synthesis rates in RT-qPCR. The Maxima™ Reverse Transcriptase is capable of cDNA synthesis at elevated temperatures (50 to 60 °C) and completion of the reverse transcription reaction in 15 to 30 minutes. The use of this enzyme in combination with Maxima™ qPCR Master Mixes ensures high specificity, sensitivity and wide linear range of two step RT-qPCR. Applications include first strand cDNA synthesis, RT-PCR, and RT-qPCR.

  • Thermostable – 90% active after incubation at 50 °C for 60 minutes in a reaction mixture
  • Active up to 60 °C
  • High yields of full-length cDNA as long as 20 kb
  • Efficient – complete cDNA synthesis in 15 to 30 minutes
  • Incorporates modified nucleotides
  • Developed through in vitro evolution

Delivery information: Supplied with 5X RT Buffer.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Maxima™ Reverse Transcriptase, 10 000 units (200 U/µl)

Specifications