Precolumn filters, ColumnSaver
PEEK, 10-32, 2,0 µm
Catalog # 55215-U
Supplier: Merck
Jump to:
Product Details & Documents
Precolumn filter kit has a standard 10-32 fitting and fits direct to most column types without introducing an additional dead volume.
Specifications
- Porosity:2,0 µm
- Thread:10-32
- Material:PEEK
- Seq:02