Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Screw top vials

Vials, with write-on patch, 4,00 ml, 15×45 mm, Amber, No, 13-425
  SIAL27116-U
 :   Merck
Screw top vials
  SIAL27116-U
 :  Merck
Screw top vials

Vials, with write-on patch, 4,00 ml, 15×45 mm, Amber, No, 13-425

 
50,20 €
   
1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
 
 
  • Cap included:
    No
  • Cap Size:
    13-425
  • Ø×H:
    15×45 mm
  • Capacity:
    4,00 ml
  • Colour:
    Amber
  • Description:
    Vials, with write-on patch
  • Seq:
    24
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.