Order Entry
Belgium
Orders LinkContactUsLinkComponent
Screw top vials

Screw top vials

Vials, 4,00 ml, 14×45 mm, Amber, No, 13-425
Catalog # SIAL27115-U
Supplier:   Merck
Screw top vials
Catalog # SIAL27115-U
Supplier:  Merck
Screw top vials

Vials, 4,00 ml, 14×45 mm, Amber, No, 13-425

Price
60,40 €
Log in for availability
1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifications
  • Cap included:
    No
  • Cap Size:
    13-425
  • Ø×H:
    14×45 mm
  • Capacity:
    4,00 ml
  • Colour:
    Amber
  • Description:
    Vials
  • Seq:
    23