Orders LinkContactUsLinkComponent
Frocks, PP
Frocks, PP
White, XL
Catalog # HOPE40.301/04
Supplier:   Medicom
Frocks, PP
Catalog # HOPE40.301/04
Supplier:  Medicom
Frocks, PP
Frocks, PP
White, XL
Price
74,50 €
Log in for availability
1 * 50 ST
Product Details & Documents
These single-use hygiene frocks made from PP protect the working environment from particles coming from the operator's body (dander, hair, perspiration, etc.).
  • With collar and long sleeves
  • Elasticated cuffs
  • Snap buttons
  • Without pockets
Weight: 40 g/m²
Specifications
  • Clothing Size:
    XL
  • Colour:
    White
  • Seq:
    03
  • Cat. No.:
    HOPE40.301/04
  • Garment application:
    General Purpose|Industrial
  • Pockets:
    Without pockets
  • Closure type:
    Press studs / snaps
  • Weight:
    40 g/m²