Orders LinkContactUsLinkComponent
Accessories for round tissue float baths, TFB
Accessories for round tissue float baths, TFB
Accessory set includes thermometer (0 - 100 °C), thermometer support and aluminium lid, Model TFB 55
Catalog # MEDT11-5500-00
Supplier:   MEDITE
Accessories for round tissue float baths, TFB
Catalog # MEDT11-5500-00
Supplier:  MEDITE
Accessories for round tissue float baths, TFB
Accessory set includes thermometer (0 - 100 °C), thermometer support and aluminium lid, Model TFB 55
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
Specifications
  • Description:
    Accessory set includes thermometer (0 - 100 °C), thermometer support and aluminium lid
  • Seq:
    08
  • For:
    Model TFB 55