Orders LinkContactUsLinkComponent
Lithium hydroxide, anhydrous 98%
Glass bottle for solids, 25 g
  13407.14
Lithium hydroxide, anhydrous 98%
  13407.14
 :  1310-65-2
Lithium hydroxide, anhydrous 98%
Lithium hydroxide, anhydrous 98%
Glass bottle for solids, 25 g
 :  1310-65-2
40,80 €
1 * 25 g
Purity: 98% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: -4+14 Mesh Density: 2.54 Appearance: White Odour: Odourless pH: 14 Melting Point: 462° Boiling Point: 924° Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095
  •  :1310-65-2
  •  :LiOH
  •  :MFCD00011095
  •  :23.95 g/mol
  •  :14,5534
  •  :924 °C
  •  :462 °C
  •  :2.54
  •  :Ambient
  •  :215-183-4
  •  :2680
  •  :8,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g