Orders LinkContactUsLinkComponent
Concentrator
Concentrator
Concentrator, solution, for use with Tube-O-DIALYZER™, 125 mL
Catalog # 786-143
Supplier:   G-Biosciences
Concentrator
Catalog # 786-143
Supplier:  G-Biosciences
CAS Number:  
Concentrator
Concentrator, solution, for use with Tube-O-DIALYZER™, 125 mL
CAS Number:  
Price
147,00 €
Log in for availability
1 * 125 mL
Product Details & Documents
The concentrator is a high molecular weight polymer which will not migrate across the dialysis membrane. The concentrator is placed against the dialysis membrane of the Tube-O-DIALYZER™, where it rapidly absorbs water from the sample and reduces the sample volume. The concentrator can be used with dialysis bags and other dialysis devices.
Specifications
Specifications
  • Pk:
    125 mL
  • Description:
    Concentrator, solution, for use with Tube-O-DIALYZER™