Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenol Red solution 0,5%

Phenol Red solution 0,5%

Catalog # 786-563
Supplier:   G-Biosciences
Phenol Red solution 0,5%
Catalog # 786-563
Supplier:  G-Biosciences
CAS Number:  143-74-8
Phenol Red solution 0,5%
CAS Number:  143-74-8
Price
55,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Phenol Red Solution [0.5%] is a pH indicator commonly used in cell culture and biochemical applications.

  • Synonym: Phenolsulfonphthalein CAS number: 143-74-8 Molecular formula: C19H14O5S Molecular weight: 354.38

At 0.5% concentration, it provides a visible color change across a physiological pH range, allowing quick assessment of pH shifts in media, buffers, or experimental reactions.

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol