Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, HyperSep™ Diol

Cartridge, Diol, 40 - 60 µm, 25 ml, 5000 mg
Catalog # HYPE60108-756
Solid phase extraction cartridges, HyperSep™ Diol
Catalog # HYPE60108-756
Solid phase extraction cartridges, HyperSep™ Diol
Solid phase extraction cartridges, HyperSep™ Diol

Cartridge, Diol, 40 - 60 µm, 25 ml, 5000 mg

Price
302,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.

  • Normal phase extraction of polar compounds
  • Purification of polar compounds

HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.

 : HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.

Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    25 ml
  • Seq:
    08
  • Bed weight:
    5000 mg
  • Phase:
    Diol