Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Brushes for Erlenmeyer flasks

Erlenmeyer piston brush, with nylon bristles, Nylon, 130 mm, 50 mm, Wire, 340 mm
  REIT10000403
Brushes for Erlenmeyer flasks
  REIT10000403
Brushes for Erlenmeyer flasks

Erlenmeyer piston brush, with nylon bristles, Nylon, 130 mm, 50 mm, Wire, 340 mm

 
71,80 €
   
1 * 10 ST
 

Brushes with natural or nylon bristles and wire handle.

 
  • Bristle material:
    Nylon
  • Brush head Ø:
    50 mm
  • Handle material:
    Wire
  • Description:
    Erlenmeyer piston brush, with nylon bristles
  • Seq:
    09
  • Total length:
    340 mm
  • Brush head length:
    130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.