Orders LinkContactUsLinkComponent
Threaded couplings
Threaded couplings
GL 45
Catalog # BOHLH900-05
Supplier:   Bohlender
Threaded couplings
Catalog # BOHLH900-05
Supplier:  Bohlender
Threaded couplings
Threaded couplings
GL 45
Price
121,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Connector, screw thread coupling, made of PTFE. Two screw caps made of PPS or PBTP with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
  • Applicable for connecting two GL threads of different sizes
  • Temperature resisitance from −200 to +250 ºC
  • Very good chemical resistance
  • FDA conformity
Specifications
  • Thread:
    GL 45