Stopcocks with GL thread, EX
8 mm, GL 25
Catalog # BOHLE712-25
Supplier: Bohlender
Jump to:
Product Details & Documents
Stopcocks with GL thread for distributing liquids or gases, quick and easy disconnection of flow.
- Cylindrical stopcock plug made of conductive PTFE-EX for good tightness
- Stop valve with mark of flow direction
- Grip made of red PP
- Suitable for pressure up to maximum 6 bar and suitable for vacuum
- Temperature resistance upto 110 °C
- Chemical resistant
Two-way stopcock made of conductive PTFE-EX with straight bore and two connections with GL thread or three-way stopcock with L-shaped or T-shaped bore and three connections with GL thread.
Specifications
- Hole Ø:8 mm
- Thread:GL 25
- Cat. No.:BOHLE712-25
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



