ContactUsLinkComponent
12.5 LITRE DIGITAL ULTRASONIC BATH, TEMP RANGE: AMBIENT+5#C to 70#C. MAINS SUPPLY: 230V AC 50/60Hz. INCLUDES STAINLESS STEEL BASKET AND ABS PLASTIC LID, 1 BOTTLE OF M2 SOLUTION, DRAIN HOSE, SD CARD,SD CARD READER AND EU CORDSET 1 * 1 items
Catalog # GRANXUB12EURO
Supplier:  GRANT INSTRUMENTS
12.5 LITRE DIGITAL ULTRASONIC BATH, TEMP RANGE: AMBIENT+5#C to 70#C. MAINS SUPPLY: 230V AC 50/60Hz. INCLUDES STAINLESS STEEL BASKET AND ABS PLASTIC LID, 1 BOTTLE OF M2 SOLUTION, DRAIN HOSE, SD CARD,SD CARD READER AND EU CORDSET 1 * 1 items
Catalog # GRANXUB12EURO
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together