Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Carbon-filtered pressure relief vent kit, incl. activated carbon filter, for ducting to outside or room; may be used with 5238500 HEPA-filtered pressure relief vent kit
Catalog # LBCP5241200
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5241200
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Carbon-filtered pressure relief vent kit, incl. activated carbon filter, for ducting to outside or room; may be used with 5238500 HEPA-filtered pressure relief vent kit
Price
2 280,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications
  • Description:
    Carbon-filtered pressure relief vent kit, incl. activated carbon filter, for ducting to outside or room; may be used with 5238500 HEPA-filtered pressure relief vent kit
  • Seq:
    12