Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum 0,2 µm filter, traps particulates; for drying train or vacuum pump exhaust; 0,25" - 0,38" ID
Catalog # LBCP5240100
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5240100
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum 0,2 µm filter, traps particulates; for drying train or vacuum pump exhaust; 0,25" - 0,38" ID
Price
562,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications
  • Description:
    Gas and vacuum 0,2 µm filter, traps particulates; for drying train or vacuum pump exhaust; 0,25" - 0,38" ID
  • Seq:
    26