Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum tubing connection kit, automatic pressure controller, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Catalog # LBCP5245200
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5245200
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum tubing connection kit, automatic pressure controller, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Price
330,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Gas and vacuum tubing connection kit, automatic pressure controller, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
  • Seq:
    03