Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Digital hygrometer, handheld, measures both percent relative humidity from 0-100% and temperature in ° F/° C; incl. 9 volt alkaline battery
Catalog # LBCP5244800
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5244800
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Digital hygrometer, handheld, measures both percent relative humidity from 0-100% and temperature in ° F/° C; incl. 9 volt alkaline battery
Price
2 440,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Digital hygrometer, handheld, measures both percent relative humidity from 0-100% and temperature in ° F/° C; incl. 9 volt alkaline battery
  • Seq:
    15