Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Pressure relief valve vent kit for exhaustig the pressure relief valve during overpressurization, incl. 3,9" OD × 3,8" ID thimble connection, gasket, two elbows, four clamps and 13" long tubing (Ductwork for outside venting and 5230600 Pressure Relief Valve are required but not included)
Catalog # LBCP5242700
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5242700
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Pressure relief valve vent kit for exhaustig the pressure relief valve during overpressurization, incl. 3,9" OD × 3,8" ID thimble connection, gasket, two elbows, four clamps and 13" long tubing (Ductwork for outside venting and 5230600 Pressure Relief Valve are required but not included)
Price
665,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Pressure relief valve vent kit for exhaustig the pressure relief valve during overpressurization, incl. 3,9" OD × 3,8" ID thimble connection, gasket, two elbows, four clamps and 13" long tubing (Ductwork for outside venting and 5230600 Pressure Relief Valve are required but not included)