Orders LinkContactUsLinkComponent
Boiling baths, SBB Aqua Plus series
SBB Aqua 26 Plus, 100 °C, 1,35/2,0 kW, 26 L, 290×145×175 mm, 390×600×445 mm
  462-0170
Boiling baths, SBB Aqua Plus series
  462-0170
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
SBB Aqua 26 Plus, 100 °C, 1,35/2,0 kW, 26 L, 290×145×175 mm, 390×600×445 mm
1 850,00 €
1 * 1 ST
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at the base of the tank, protected by a perforated tray, giving more usable space
  • Robust construction, durable and resistant to corrosion
  • New modern design
Required accessories must be ordered separately, see SUB Aqua Plus baths entry.
 : Required accessories must be ordered separately, see SUB Aqua Pro baths entry.
 : Supplied with polycarbonate lid but without rings.
 : ** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
  • Heating capacity:
    1,35/2,0 kW
  • Temp.:
    100 °C
  • Capacity:
    26 L
  • Seq:
    04
  • W×D×H:
    390×600×445 mm
  • Bath W×D×H:
    290×145×175 mm
  • Type:
    SBB Aqua 26 Plus
Accessories
  •  
    Description
    For
  • 462-8458
    Stainless steel rack for 1.5 ml microtubes
    65 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8457
    Stainless steel rack for 0.5 ml microtubes
    105 tubes
    1 * 1 ST - 152,00 €
  •  
    Description
    For
  • 462-8350
    Stainless steel base tray**
    SAP12, JBN12, JBA12, GLS Aqua 12 Plus , SBB Aqua 12 Plus
    1 * 1 ST - 116,00 €
  •  
    Description
    For
  • 462-8351
    Stainless steel base tray**
    SAP18/26. JBN18/26, JBA18, GLS Aqua 18 Plus, OLS200 , SBB Aqua 18/26 Plus
    1 * 1 ST - 123,00 €
  •  
    Description
    For
  • 462-8456
    Stainless steel test tube rack for 30 mm Ø tubes
    12 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8454
    Stainless steel test tube rack for 19 mm Ø tubes
    32 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8455
    Stainless steel test tube rack for 25 mm Ø tubes
    18 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8453
    Stainless steel test tube rack for 16 mm Ø tubes
    36 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8452
    Stainless steel test tube rack for 13 mm Ø tubes
    55 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8451
    Stainless steel test tube rack for 10 mm Ø tubes
    84 tubes
    1 * 1 ST - 150,00 €
  •  
    Description
    For
  • 462-8212
    Gabled stainless steel lid
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26, GLS Aqua 18 Plus), LSB18
    1 * 1 ST - 296,00 €
  •  
    Description
    For
  • 462-8211
    Gabled stainless steel lid
    JBA12, JBN12, SAP12, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus, GLS Aqua 12 Plus), LSB12
    1 * 1 ST - 264,00 €
  •  
    Description
    For
  • 462-8210
    Gabled stainless steel lid
    JBA5, JBN5, SAP2S, SAP5, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    1 * 1 ST - 240,00 €
  •  
    Description
    For
  • 462-8222
    Flat lid with 6 ring sets
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26 Plus)
    1 * 1 ST - 611,00 €
  •  
    Description
    For
  • 462-8221
    Flat lid with 4 ring sets
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    1 * 1 ST - 440,00 €
  •  
    Description
    For
  • 462-8232
    Raised half shelf with perforated mesh
    JBN26, SAP26, (SUB Aqua 26, JB Aqua 26, SBB Aqua 26 Plus)
    1 * 1 ST - 255,00 €
  •  
    Description
    For
  • 462-8220
    Flat lid with 2 ring sets
    JBA5, JBN5, SAP5, SAPD, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    1 * 1 ST - 286,00 €
  •  
    Description
    For
  • 462-0187
    Raised half shelf with perforated mesh
    JBA18, JBN18, SAP18, (SUB Aqua 18, JB Aqua 18, SBB Aqua 18 Plus)
    1 * 1 ST - 215,00 €
  •  
    Description
    For
  • 462-8231
    Raised half shelf with perforated mesh
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    1 * 1 ST - 182,00 €
  •  
    Description
    For
  • GRANAQL5
    Replacement lids, polycarbonate
    SAP2S, SAP5 , JBN5, JBA5, SBB Aqua 5 Plus
    1 * 1 ST - 122,00 €
  •  
    Description
  • 462-0333
    Polycarbonate lid
    1 * 1 ST - 150,00 €
  •  
    Description
  • 462-0334
    Polycarbonate lid
    1 * 1 ST - 171,00 €
  •  
    Description
  • 462-8131
    Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    1 * 1 ST - 123,00 €
  •  
    Description
    Material
    Ø
  • 462-8239
    Anti-evaporation spheres
    PP
    20 mm
    1 * 300 ST - 123,00 €