ContactUsLinkComponent
SBB Aqua 18 Plus, 18 litre unstirred boiling water bath. Temp range: 100C only. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cordset 1 * 1 items
N° de catalogue 462-0167
Fournisseur:  GRANT INSTRUMENTS
SBB Aqua 18 Plus, 18 litre unstirred boiling water bath. Temp range: 100C only. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cordset 1 * 1 items
N° de catalogue 462-0167
Fournisseur:  GRANT INSTRUMENTS
Frequently Bought Together