STANDARD VISCOSITY N2000 60 TO 135°C 1 * 500 ml
Catalog # CANN9727-E15
Supplier: CANNON INSTRUMENT
Frequently Bought Together
BRILLIANT BLACK BN 1 * 0,25 g
Supplier: EHRENSTORFER
PARAQUAT DICHLORIDE D8 1 * 50 mg
Supplier: EHRENSTORFER
TETRABROMOBISPHENOL A-DIGLYCIDYL ETHER 1 * 25 mg
Supplier: EHRENSTORFER
Gold standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
CLOUD POINT APPROX. VALUE (-20 C) 1 * 250 ml
Supplier: AccuStandard
Supelco® Organic Reference Standard, Hexanal
Supplier: Merck
Coumarin
Supplier: EHRENSTORFER
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
BUFFER SOLUTION PH=4,01 1 * 5 l
Supplier: Mettler - Toledo
4-Hydroxy-3,5-diiodobenzonitrile
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, α-Arbutin
Supplier: Merck