ContactUsLinkComponent
Stirring-heating block combination with 15 stirring positions, 15 bore holes, # 71,5 mm, depth 78 mm, distance 84 mm, suitable for 2 50 ml lab-flasks (e.g. Duran) and for 400 ml beaker glasses incl. control unit. 1 * 1 items
Catalog # 460-0138
Supplier:  2MAG
Stirring-heating block combination with 15 stirring positions, 15 bore holes, # 71,5 mm, depth 78 mm, distance 84 mm, suitable for 2 50 ml lab-flasks (e.g. Duran) and for 400 ml beaker glasses incl. control unit. 1 * 1 items
Catalog # 460-0138
Supplier:  2MAG
Frequently Bought Together